L(+)-Citrulline 99%
Supplier: Thermo Scientific
110470250
110471000
ACRO110470250EA
177
EUR
ACRO110470250
ACRO110471000
L(+)-Citrulline 99%
L(+)-Citrulline
Formula:
C₆H₁₃N₃O₃ Melting Pt: 214 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064397 CAS Number: 372-75-8 EINECS: 206-759-6 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.3 % (1 g, 105°C, 3 hrs) |
Residue after ignition | ≤0.1 % |
UV transmittance | ≥95.0 % (c=10, H2O, 430 nm) 1 cm cell |
Specific optical rotation | +24.5° to +26.8° (20°C, 589 nm) (c=8, 6N HCl) |
Titration with HClO4 | 98.5 to 101.0 % |
Chloride (Cl) | ≤200 ppm |
Sulfate (SO4) | ≤200 ppm |
Ammonium (NH4) | ≤200 ppm |
Arsenic (As) | ≤1 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...