Deoxybenzoin 98%
Supplier: Thermo Scientific
Synonyms:
2-Phenylacetophenone, 1,2-Diphenylethanone
111960250
111961000
ACRO111960250EA
42.3
EUR
ACRO111960250
ACRO111961000
Deoxybenzoin 98%
Deoxybenzoin
Formula:
C₁₄H₁₂O MW: 196,25 g/mol Melting Pt: 52…57 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003081 CAS Number: 451-40-1 EINECS: 207-193-2 |
Specification Test Results
Appearance | White to yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 52 to 57 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...