4-Methylphenethylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(p-Tolyl)ethylamine
157880010
157880050
ACRO157880010EA
38.7
EUR
ACRO157880010
ACRO157880050
4-Methylphenethylamine 97%
4-Methylphenethylamine
Formula:
C₉H₁₃N MW: 135,21 g/mol Boiling Pt: 214 °C (760 mmHg) Density: 0,93 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008195 CAS Number: 3261-62-9 EINECS: 221-865-2 UN: 2922 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5250 to 1.5280 (20°C, 589 nm) |
GC | ≥96.0 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...