Triisobutylaluminium 1.1 M toluene, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TIBAL, Tri-i-butylaluminum

185421000 185428000
185421000.EA 102 EUR
185421000. 185428000.
Triisobutylaluminium 1.1 M toluene, AcroSeal®
Triisobutylaluminium
Formula: C₁₂H₂₇Al
MW: 198.33 g/mol
Density: 0.84 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008929
CAS Number: 100-99-2
EINECS: 202-906-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Assay 24.5 to 25.5 % (on Al basis)
Loss on drying ≤8 %
Solubility (5 % in Ethanol, hot) Clear to hazy
HPLC ≥96.0 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR