Triethylaluminium 0.6 M heptane, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Triethylaluminum

381171000 381178000
381171000.EA 110 EUR
381171000. 381178000.
Triethylaluminium 0.6 M heptane, AcroSeal®
Triethylaluminium
Formula: C₆H₁₅Al
MW: 114.17 g/mol
Boiling Pt: 128
Melting Pt: -46.1
Storage Temperature: Ambient
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 9.5 to 10.5 wt % (on Al basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR