Desyl bromide 97%

Supplier: Thermo Fisher Scientific
Danger

403170250 403170050
ACRO403170250EA 307 EUR
ACRO403170250 ACRO403170050
Desyl bromide 97%
Desyl bromide
Formula: C₁₄H₁₁BrO
Melting Pt: 53…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000136
CAS Number: 1484-50-0
EINECS: 216-057-1
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR