Deoxybenzoin ≥97%
Supplier: Thermo Fisher Scientific
A10580.14
A10580.22
A10580.14EA
35
EUR
A10580.14
A10580.22
Deoxybenzoin ≥97%
Deoxybenzoin
Purity: 97%
CAS No.: 451-40-1
Molecular Formula: C14H12O
Formula Weight: 196.25
Flash Point:
Formula:
C₁₄H₁₂O Boiling Pt: 319…321 °C Melting Pt: 53…57 °C Density: 1,2 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003081 CAS Number: 451-40-1 EINECS: 207-193-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...