2-Chloroethylammonium chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride
A14455.30
A14455.0B
A14455.18
A14455.30EA
118
EUR
A14455.30
A14455.0B
A14455.18
2-Chloroethylammonium chloride ≥98%
2-Chloroethylammonium chloride
Purity: 98+%
CAS No.: 870-24-6
Molecular Formula: ClCH2CH2NH2.HCl
Formula Weight: 115.99
Odour: Odourless
pH: 2-3
Melting Point: 143-148°
Beilstein Reference: 3590304
UN No.: 3261
MDL No.: MFCD00012887
Formula:
C₂H₇Cl₂N MW: 115.99 g/mol Melting Pt: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012887 CAS Number: 870-24-6 EINECS: 212-793-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...