4-Morpholinoaniline ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-(4-Morpholinyl)aniline
B20292.06
B20292.14
B20292.03
B20292.06EA
48.3
EUR
B20292.06
B20292.14
B20292.03
4-Morpholinoaniline ≥98%
4-Morpholinoaniline
Purity: 98+%
CAS No.: 2524-67-6
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 129-133°
Beilstein Reference: 150957
UN No.: 2811
MDL No.: MFCD00006169
Formula:
C₁₀H₁₄N₂O MW: 178,23 g/mol Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006169 CAS Number: 2524-67-6 EINECS: 219-760-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...