N-(2-Hydroxyethyl)morpholine ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
B22759.22
B22759.30
B22759.36
B22759.22EA
30.8
EUR
B22759.22
B22759.30
B22759.36
N-(2-Hydroxyethyl)morpholine ≥99%
N-(2-Hydroxyethyl)morpholine
Purity: 99%
CAS No.: 622-40-2
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Flash Point: 99°(210°F)
Refractive Index: 1.4775
Melting Point: 1-2°
Boiling Point: 226-228°
Beilstein Reference: 104375
MDL No.: MFCD00006180
Formula:
C₆H₁₃NO₂ Boiling Pt: 226…228 °C Melting Pt: 1…2 °C Density: 1,080 Flash Pt: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006180 CAS Number: 622-40-2 EINECS: 210-734-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...