4-(4-Nitrophenyl)morpholine ≥98%

Supplier: Thermo Scientific
Warning

B24933.06 B24933.14 B24933.22
B24933.06EA 26.5 EUR
B24933.06 B24933.14 B24933.22
4-(4-Nitrophenyl)morpholine ≥98%
4-(4-Nitrophenyl)morpholine

Purity: 98% CAS No.: 10389-51-2 Molecular Formula: C10H12N2O3 Formula Weight: 208.22 Melting Point: 148-152° MDL No.: MFCD00023317

Formula: C₁₀H₁₂N₂O₃
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00023317
CAS Number: 10389-51-2
EINECS: 233-851-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR