5-Formyl-2-thiopheneboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid
H29289.06
H29289.03
H29289.06EA
305
EUR
H29289.06
H29289.03
5-Formyl-2-thiopheneboronic acid ≥97%
5-Formyl-2-thiopheneboronic acid
Purity: 97%
CAS No.: 4347-33-5
Molecular Formula: C5H5BO3S
Formula Weight: 155.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 124° dec.
MDL No.: MFCD02093666
Formula:
C₅H₅BO₃S MW: 155,97 g/mol Melting Pt: ∼ 124 °C (dec.) |
MDL Number:
MFCD02093666 CAS Number: 4347-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...