3-Methylbenzamidoxime 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
H50682.03
H50682.06
H50682.03EA
0
EUR
H50682.03
H50682.06
3-Methylbenzamidoxime 96%
3-Methylbenzamidoxime
Purity: 96%
CAS No.: 40067-82-1
Molecular Formula: C8H10N2O
Formula Weight: 150.18
Melting Point: 87-90°
MDL No.: MFCD00655170
Formula:
C₈H₁₀N₂O Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833266 CAS Number: 40067-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...