1-(2-Bromoethoxy)-3,5-dimethylbenzene ≥97%

Supplier: Thermo Scientific
Warning

H52397.MD
H52397.MDEA 43.4 EUR
H52397.MD
1-(2-Bromoethoxy)-3,5-dimethylbenzene ≥97%
1-(2-Bromoethoxy)-3,5-dimethylbenzene

Purity: 97+% CAS No.: 37136-93-9 Molecular Formula: C10H13BrO Formula Weight: 229.12 MDL No.: MFCD02030859

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD02030859
CAS Number: 37136-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR