Ethanolamine hydrochloride ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride
L14322.22
L14322.36
L14322.22EA
37.1
EUR
L14322.22
L14322.36
Ethanolamine hydrochloride ≥98%
Ethanolamine hydrochloride
Purity: 98+%
CAS No.: 2002-24-6
Molecular Formula: HOCH2CH2NH2.HCl
Formula Weight: 97.55
Melting Point: 83-86°
Beilstein Reference: 3542892
Merck Reference: 14,3727
MDL No.: MFCD00012892
Formula:
C₂H₈ClNO MW: 97,54 g/mol Melting Pt: 83…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012892 CAS Number: 2002-24-6 EINECS: 217-900-6 Merck Index: 14,3727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...