3-Methylthiophene-2-carbonyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
L17989.09
L17989.04
L17989.09EA
136
EUR
L17989.09
L17989.04
3-Methylthiophene-2-carbonyl chloride ≥98%
3-Methylthiophene-2-carbonyl chloride
Purity: 98%
CAS No.: 61341-26-2
Molecular Formula: C6H5ClOS
Formula Weight: 160.62
Refractive Index: 1.5855
Boiling Point: 216-220°
UN No.: 3265
MDL No.: MFCD00130091
Formula:
C₆H₅ClOS MW: 160,62 g/mol Boiling Pt: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130091 CAS Number: 61341-26-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...