Pentyl nitrite (mixed isomers ), Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Amyl Nitrite, Pentyl Nitrite
M-Clarity™ quality level = MQ200
8.20966.0005
8.20966.0250
8.20966.0005EA
31.7
EUR
8.20966.0005
8.20966.0250
Pentyl nitrite (mixed isomers ), Sigma-Aldrich®
Pentyl nitrite (mixed isomers )
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036132 CAS Number: 463-04-7 EINECS: 207-332-7 UN: 1113 ADR: 3 II Merck Index: 12,01617 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Molar Mass | 117.14 g/mol |
Boiling Point | 104 °C (1013 hPa) |
Vapor Pressure | 65 hPa (20 °C) |
Flash Point | -40 °C |
Density | 0.86 g/cm3 (20 °C) |
Ignition Point | 205 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...