3,5-Xylidine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3,5-Dimethylaniline, 5-Amino-m-xylene

A10570.22 A10570.36
A10570.22EA 51 EUR
A10570.22 A10570.36
3,5-Xylidine ≥97%
3,5-Xylidine

Purity: 97+% CAS No.: 108-69-0 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 93°(199°F) Refractive Index: 1.5580 Odour: Characteristic Melting Point: 9-11° Boiling Point: 220-221° Beilstein Reference: 507051 Merck Reference: 14,10084 UN No.: 1711 MDL No.: MFCD00007813

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 220…221 °C
Melting Pt: 9…11 °C
Density: 0.971
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1 II
Merck Index: 12,10217

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR