1,4-Di-tert-butylbenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A17146.06
A17146.14
A17146.06EA
56.7
EUR
A17146.06
A17146.14
1,4-Di-tert-butylbenzene ≥98%
1,4-Di-tert-butylbenzene
Purity: 98%
CAS No.: 1012-72-2
Molecular Formula: C14H22
Formula Weight: 190.33
Melting Point: 75-79°
Boiling Point: 236°
MDL No.: MFCD00008836
Formula:
C₁₄H₂₂ MW: 190.33 g/mol Boiling Pt: 236 °C Melting Pt: 75…79 °C Density: 0.985 Storage Temperature: Ambient |
MDL Number:
MFCD00008836 CAS Number: 1012-72-2 EINECS: 213-790-9 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...