2,4,6-Triisopropylbenzoic acid ≥97%

Supplier: Thermo Scientific

A15489.09 A15489.18
A15489.09EA 70 EUR
A15489.09 A15489.18
2,4,6-Triisopropylbenzoic acid ≥97%
2,4,6-Triisopropylbenzoic acid

Purity: 97% CAS No.: 49623-71-4 Molecular Formula: C16H24O2 Formula Weight: 248.37 Melting Point: 184-187° Beilstein Reference: 2372331

Formula: C₁₆H₂₄O₂
MW: 248,37 g/mol
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00015031
CAS Number: 49623-71-4
EINECS: 256-400-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR