2,3,5-Trimethylphenol ≥98%
Supplier: Thermo Scientific
|
Danger
|
A10745.0B
A10745.14
A10745.22
A10745.0BEA
581
EUR
A10745.0B
A10745.14
A10745.22
2,3,5-Trimethylphenol ≥98%
2,3,5-Trimethylphenol
Purity: 98+%
CAS No.: 697-82-5
Molecular Formula: C9H12O
Formula Weight: 136.20
Odour: Phenol-like
Melting Point: 92-95°
Boiling Point: 230-231°
Beilstein Reference: 2042210
UN No.: 2430
MDL No.: MFCD00002228
Formula:
C₉H₁₂O MW: 136,19 g/mol Boiling Pt: 230…231 °C Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002228 CAS Number: 697-82-5 EINECS: 211-806-9 UN: 2430 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...