1,3,5-Triisopropylbenzene 95%
Supplier: Thermo Scientific
171221000
171220250
ACRO171221000EA
108
EUR
ACRO171221000
ACRO171220250
1,3,5-Triisopropylbenzene 95%
1,3,5-Triisopropylbenzene
Formula:
C₁₅H₂₄ MW: 204,36 g/mol Boiling Pt: 232…236 °C (760 mmHg) Melting Pt: -7 °C Density: 0,84 g/cm³ Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008890 CAS Number: 717-74-8 EINECS: 211-941-3 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4874 to 1.4894 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...