1,3,5-Triisopropylbenzene 95%
Supplier: Thermo Scientific
Warning
|
A13453.0B
A13453.18
A13453.30
A13453.0BEA
604
EUR
A13453.0B
A13453.18
A13453.30
1,3,5-Triisopropylbenzene 95%
1,3,5-Triisopropylbenzene
Purity: 95%
CAS No.: 717-74-8
Molecular Formula: C15H24
Formula Weight: 204.36
Flash Point: 86°(186°F)
Refractive Index: 1.4880
Melting Point: -7°
Boiling Point: 235-237°
Beilstein Reference: 1862750
MDL No.: MFCD00008890
Formula:
C₁₅H₂₄ MW: 204,36 g/mol Boiling Pt: 235…237 °C Melting Pt: –7 °C Density: 0,854 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008890 CAS Number: 717-74-8 EINECS: 211-941-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...