1,2,4,5-Tetramethylbenzene ≥97%

Supplier: Thermo Scientific
Warning

L04493.22 L04493.36
L04493.22EA 33.9 EUR
L04493.22 L04493.36
1,2,4,5-Tetramethylbenzene ≥97%
1,2,4,5-Tetramethylbenzene

Purity: 97+% CAS No.: 95-93-2 Molecular Formula: C10H14 Formula Weight: 134.22 Flash Point: 73°(163°F) Melting Point: 77-80° Boiling Point: 191-193° Beilstein Reference: 1903393 Merck Reference: 14,3468 UN No.: 1325 MDL No.: MFCD00008528

Formula: C₁₀H₁₄
MW: 134,22 g/mol
Boiling Pt: 191…193 °C
Melting Pt: 77…80 °C
Density: 1,036 g/cm³ (20 °C)
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008528
CAS Number: 95-93-2
EINECS: 202-465-7
UN: 1325
ADR: 4.1,III
Merck Index: 14,3468

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR