4-Phenylcyclohexanone 98+%
Supplier: Thermo Fisher Scientific
Synonyms:
4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane
187940500
187940100
ACRO187940500EA
145
EUR
ACRO187940500
ACRO187940100
4-Phenylcyclohexanone 98+%
4-Phenylcyclohexanone
Formula:
C₁₂H₁₄O Boiling Pt: 294 °C (1013 hPa) Melting Pt: 77…80 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001641 CAS Number: 4894-75-1 EINECS: 225-517-0 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 76 to 80 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...