6-Quinolinecarboxylic acid 98%

Supplier: Thermo Scientific
Warning

418800050 418800250
ACRO418800050EA 96.7 EUR
ACRO418800050 ACRO418800250
6-Quinolinecarboxylic acid 98%
6-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173,17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 293…294 °C
Storage Temperature: Ambient
MDL Number: MFCD00047613
CAS Number: 10349-57-2
EINECS: 233-761-4

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR