8-Aminoquinoline 98+%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Quinolin-8-ylamine
123710050
123710100
123710500
ACRO123710050EA
102
EUR
ACRO123710050
ACRO123710100
ACRO123710500
8-Aminoquinoline 98+%
8-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 174 °C (26 mmHg) Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006809 CAS Number: 578-66-5 EINECS: 209-427-9 UN: 2811 ADR: 6.1 III |
Specification Test Results
Appearance | Beige to green to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 62 to 65 °C |
GC | ≥98.0 % |
Titration with HClO4 | ≥98.0 % |
Aluminium (Al) | ≤0.000005 % |
Barium (Ba) | ≤0.000002 % |
Bismuth (Bi) | ≤0.00001 % |
Cadmium (Cd) | ≤0.000001 % |
Calcium (Ca) | ≤0.00005 % |
Chromium (Cr) | ≤0.000002 % |
Cobalt (Co) | ≤0.000001 % |
Copper (Cu) | ≤0.000002 % |
Iron (Fe) | ≤0.00002 % |
Lead (Pb) | ≤0.000002 % |
Lithium (Li) | ≤0.000001 % |
Magnesium (Mg) | ≤0.00001 % |
Manganese (Mn) | ≤0.000001 % |
Molybdenum (Mo) | ≤0.000002 % |
Nickel (Ni) | ≤0.00002 % |
Potassium (K) | ≤0.00001 % |
Sodium (Na) | ≤0.00005 % |
Titanium (Ti) | ≤0.00001 % |
Vanadium (V) | ≤0.000001 % |
Zirconium (Zr) | ≤0.00001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...