8-Quinolinesulphonic acid ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 8-Quinolinesulphonic acid

L06640.09 L06640.18
L06640.09EA 68.8 EUR
L06640.09 L06640.18
8-Quinolinesulphonic acid ≥98%
8-Quinolinesulphonic acid

Purity: 98% CAS No.: 85-48-3 Molecular Formula: C9H7NO3S Formula Weight: 209.23 Melting Point: 300° Beilstein Reference: 9501 UN No.: 2585 MDL No.: MFCD00024037

Formula: C₉H₇NO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00024037
CAS Number: 85-48-3
EINECS: 201-611-7
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR