4-Chloroquinaldine ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15002.03
A15002.06
A15002.14
A15002.03EA
40.5
EUR
A15002.03
A15002.06
A15002.14
4-Chloroquinaldine ≥97%
4-Chloroquinaldine
Purity: 97%
CAS No.: 4295-06-1
Molecular Formula: C10H8ClN
Formula Weight: 177.63
Flash Point:
Formula:
C₁₀H₈ClN MW: 177.63 g/mol Boiling Pt: 269…270 °C Melting Pt: ∼25 °C Density: 0.881 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006757 CAS Number: 4295-06-1 EINECS: 224-300-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...