6-Nitroquinaldine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A10945.06
A10945.03
A10945.14
A10945.06EA
160
EUR
A10945.06
A10945.03
A10945.14
6-Nitroquinaldine ≥98%
6-Nitroquinaldine
Purity: 98%
CAS No.: 613-30-9
Molecular Formula: C10H8N2O2
Formula Weight: 188.19
Melting Point: 162-166°
Beilstein Reference: 7915
Formula:
C₁₀H₈N₂O₂ Melting Pt: 162…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051736 CAS Number: 613-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...