8-Nitroquinaldine ≥98%

Supplier: Thermo Fisher Scientific
Warning

L03006.03 L03006.06
L03006.03EA 15.3 EUR
L03006.03 L03006.06
8-Nitroquinaldine ≥98%
8-Nitroquinaldine

Purity: 98% CAS No.: 881-07-2 Molecular Formula: C10H8N2O2 Formula Weight: 188.19 Odour: Odourless Melting Point: 136-139° Beilstein Reference: 145077 MDL No.: MFCD00006764

Formula: C₁₀H₈N₂O₂
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006764
CAS Number: 881-07-2
EINECS: 212-919-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR