6-Methoxyquinaldine 97
Supplier: Thermo Fisher Scientific
|
Warning
|
B22199.03
B22199.06
B22199.03EA
45.6
EUR
B22199.03
B22199.06
6-Methoxyquinaldine 97
6-Methoxyquinaldine
Purity: 97%
CAS No.: 1078-28-0
Molecular Formula: C11H11NO
Formula Weight: 173.21
Flash Point: 47°(117°F)
Melting Point: 62-66°
MDL No.: MFCD00006761
Formula:
C₁₁H₁₁NO MW: 173.21 g/mol Boiling Pt: 95…96 °C (0.5 mmHg) Melting Pt: 62…66 °C Density: 0.973 Flash Pt: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006761 CAS Number: 1078-28-0 EINECS: 214-080-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...