Cyclopentanone 99
Supplier: Thermo Fisher Scientific
|
Warning
|
A14222.0F
A14222.AK
A14222.AP
A14222.0FEA
192
EUR
A14222.0F
A14222.AK
A14222.AP
Cyclopentanone 99
Cyclopentanone
Purity: 99%
CAS No.: 120-92-3
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 30°(86°F)
Refractive Index: 1.4370
Odour: Petrol-like
Melting Point: -51°
Boiling Point: 130-131°
Beilstein Reference: 605573
Merck Reference: 14,2743
UN No.: 2245
MDL No.: MFCD00001409
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 130…131 °C Melting Pt: –51 °C Density: 0.949 Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001409 CAS Number: 120-92-3 EINECS: 204-435-9 UN: 2245 ADR: 3 III Merck Index: 13,02771 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...