Cyclopropanecarboxylic acid ≥98%

Supplier: Thermo Scientific
Danger

A14215.0B A14215.18 A14215.30
A14215.0BEA 509 EUR
A14215.0B A14215.18 A14215.30
Cyclopropanecarboxylic acid ≥98%
Cyclopropanecarboxylic acid

Purity: 98% CAS No.: 1759-53-1 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 71°(159°F) Refractive Index: 1.4390 Melting Point: 5° Boiling Point: 182-184° Beilstein Reference: 969839 UN No.: 3265 MDL No.: MFCD00001287

Formula: C₄H₆O₂
MW: 86,09 g/mol
Boiling Pt: 182…184 °C
Melting Pt: 5 °C
Density: 1,086
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001287
CAS Number: 1759-53-1
EINECS: 217-162-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR