Chlorocyclohexane ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cyclohexyl chloride

L04349.30 L04349.0B
L04349.30EA 42.5 EUR
L04349.30 L04349.0B
Chlorocyclohexane ≥98%
Chlorocyclohexane

Purity: 98% CAS No.: 542-18-7 Molecular Formula: C6H11Cl Formula Weight: 118.61 Flash Point: 28°(82°F) Refractive Index: 1.4620 Odour: Unpleasant Melting Point: -44° Boiling Point: 141-143° Beilstein Reference: 1900796 Merck Reference: 14,2732 UN No.: 1993 MDL No.: MFCD00003822

Formula: C₆H₁₁Cl
Boiling Pt: 141…143 °C
Melting Pt: –44 °C
Density: 1,000
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003822
CAS Number: 542-18-7
EINECS: 208-806-6
UN: 1993
ADR: 3,III
Merck Index: 14,2732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR