Cyclooctylamine 97%

Supplier: Thermo Fisher Scientific
Danger

111460050 111460250 111461000
ACRO111460050EA 33.7 EUR
ACRO111460050 ACRO111460250 ACRO111461000
Cyclooctylamine 97%
Cyclooctylamine
Formula: C₈H₁₇N
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: –48 °C
Density: 0,92 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001748
CAS Number: 5452-37-9
EINECS: 226-694-7
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR