Cyclobutanol 96%
Supplier: Thermo Scientific
|
Danger
|
A12430.03
A12430.06
A12430.14
A12430.03EA
151
EUR
A12430.03
A12430.06
A12430.14
Cyclobutanol 96%
Cyclobutanol
Purity: 96%
CAS No.: 2919-23-5
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: 21°(69°F)
Refractive Index: 1.4360
Boiling Point: 123-124°
Beilstein Reference: 2035937
UN No.: 1120
MDL No.: MFCD00001318
Formula:
C₄H₈O MW: 72,11 g/mol Boiling Pt: 123…124 °C Density: 0,925 Flash Pt: 21 °C (69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001318 CAS Number: 2919-23-5 EINECS: 220-858-1 UN: 1120 ADR: 3,II |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...