Cyclohexanol 98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Hydroxycyclohexane, Hexahydrophenol

147680010 147680025 147680100
ACRO147680010EA 67.8 EUR
ACRO147680010 ACRO147680025 ACRO147680100
Cyclohexanol 98%
Cyclohexanol
Formula: C₆H₁₂O
MW: 100,16 g/mol
Boiling Pt: 161 °C (760 mmHg)
Melting Pt: 23 °C
Density: 0,96 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8878907005758628f5927d7b0f43a99f57de5ae8&vid=3e9ba5e70ee55ec56d3b4ee09842971154838cc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4670 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR