Cycloheptane 99%

Supplier: Thermo Fisher Scientific
Danger

111010050 111011000 111010250
ACRO111010050EA 41 EUR
ACRO111010050 ACRO111011000 ACRO111010250
Cycloheptane 99%
Cycloheptane
Formula: C₇H₁₄
Boiling Pt: 118,4 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0,81 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004147
CAS Number: 291-64-5
EINECS: 206-030-2
UN: 2241
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
Acid value 230 to 260 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR