2-Chloroethyldiethylammonium chloride 99.5%
Supplier: Thermo Fisher Scientific
|
Danger
|
114030010
114030050
114032500
114030025
ACRO114030010EA
128
EUR
ACRO114030010
ACRO114030050
ACRO114032500
ACRO114030025
2-Chloroethyldiethylammonium chloride 99.5%
2-Chloroethyldiethylammonium chloride
Formula:
C₆H₁₅Cl₂N Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012519 CAS Number: 869-24-9 EINECS: 212-786-4 UN: 2928 ADR: 6.1,II |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 208 to 212 °C |
Loss on drying | ≤1 % (105 °C, 3 hrs) |
Titration with HClO4 | ≥99.4 % (On dry substance) |
Zinc (Zn) | ≤50 ppm |
Heavy metals (as Pb) | ≤30 ppm |
Sugars, dextrin | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...