2-Furoic acid 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Furan-2-carboxylic acid, 2-Furancarboxylic acid
119820050
119821000
119825000
119820025
ACRO119820050EA
26.9
EUR
ACRO119820050
ACRO119821000
ACRO119825000
ACRO119820025
2-Furoic acid 98%
2-Furoic acid
Formula:
C₅H₄O₃ MW: 112,08 g/mol Boiling Pt: 230…232 °C (760 mmHg) Melting Pt: 129…133 °C Flash Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003238 CAS Number: 88-14-2 EINECS: 201-803-0 |
Specification Test Results
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 129 to 133 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...