(-)-Menthone 85%

Supplier: Thermo Fisher Scientific
Warning

203610250 203611000
ACRO203610250EA 55 EUR
ACRO203610250 ACRO203611000
(-)-Menthone 85%
(-)-Menthone
Formula: C₁₀H₁₈O
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00001634
CAS Number: 14073-97-3
EINECS: 237-926-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4530 (20°C, 589 nm)
GC ≥80 %
Specific optical rotation -2° to -10° (20°C, 589 nm) (c=4, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR