Zinc trifluoromethanesulphonate 98%

Supplier: Thermo Scientific
Danger

317090100 317090500
ACRO317090100EA 90.9 EUR
ACRO317090100 ACRO317090500
Zinc trifluoromethanesulphonate 98%
Zinc trifluoromethanesulphonate
Formula: C₂F₆O₆S₂Zn
MW: 363,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013229
CAS Number: 54010-75-2
EINECS: 258-922-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light grey Powder
Infrared spectrum Conforms
Titration Complexometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR