Bis(2-ethylhexyl)adipate 99%

Supplier: Thermo Scientific
Warning

Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

402460010 402462500 402460025
ACRO402460010EA 46.8 EUR
ACRO402460010 ACRO402462500 ACRO402460025
Bis(2-ethylhexyl)adipate 99%
Bis(2-ethylhexyl) adipate
Formula: C₂₂H₄₂O₄
MW: 370,57 g/mol
Boiling Pt: 210…218 °C (5 mmHg)
Melting Pt: –76 °C
Density: 0,924 g/cm³
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4500 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤30 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR