Didecyl phthalate 95%
Supplier: Thermo Fisher Scientific
407100250
407101000
ACRO407100250EA
90.2
EUR
ACRO407100250
ACRO407101000
Didecyl phthalate 95%
Didecyl phthalate
Formula:
C₂₈H₄₆O₄ Boiling Pt: 261 °C (5 mmHg) Melting Pt: 4 °C Flash Pt: 232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041915 CAS Number: 84-77-5 EINECS: 201-561-6 |
Specification Test Results
Appearance | Clear to hazy yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4780 to 1.4840 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...