Ethyl-3-bromopropionate 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
409811000
409810250
ACRO409811000EA
168
EUR
ACRO409811000
ACRO409810250
Ethyl-3-bromopropionate 99%
Formula:
C₅H₉BrO₂ Boiling Pt: 69…71 °C (10 mmHg) Density: 1,419 g/cm³ Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000251 CAS Number: 539-74-2 EINECS: 208-724-0 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4540 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...