2-Furoic acid ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid

A11454.22 A11454.36
A11454.22EA 35.3 EUR
A11454.22 A11454.36
2-Furoic acid ≥98%
2-Furoic acid

Purity: 98% CAS No.: 88-14-2 Molecular Formula: C5H4O3 Formula Weight: 112.08 Flash Point: 137°(278°F) Odour: Odourless Boiling Point: 230-232° subl. Beilstein Reference: 110149 Merck Reference: 14,4307 MDL No.: MFCD00003238

Formula: C₅H₄O₃
MW: 112,08 g/mol
Boiling Pt: 230…232 °C (subl.)
Melting Pt: 127…132 °C
Flash Pt: 137 °C (278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003238
CAS Number: 88-14-2
EINECS: 201-803-0
Merck Index: 14,4307

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR