1-Formylpiperazine ≥90%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Piperazinecarboxaldehyde, 1-Piperazinecarbaldehyde

A14757.14 A14757.22 A14757.36
A14757.14EA 80.7 EUR
A14757.14 A14757.22 A14757.36
1-Formylpiperazine ≥90%
1-Formylpiperazine

Purity: 90+% CAS No.: 7755-92-2 Molecular Formula: C5H10N2O Formula Weight: 114.15 Storage Temperature: 2°C to 8°C Flash Point: 102°(216°F) Refractive Index: 1.5120 Boiling Point: 264-266° Beilstein Reference: 110295 MDL No.: MFCD00005963

Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 264…266 °C
Density: 1.111
Flash Pt: 102 °C (216 °F)
MDL Number: MFCD00005963
CAS Number: 7755-92-2
EINECS: 231-813-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR