Trimethyloxonium tetrafluoroborate 96%
Supplier: Thermo Scientific
|
Danger
|
A15175.09
A15175.18
A15175.30
A15175.09EA
111
EUR
A15175.09
A15175.18
A15175.30
Trimethyloxonium tetrafluoroborate 96%
Trimethyloxonium tetrafluoroborate
Purity: 96%
CAS No.: 420-37-1
Molecular Formula: C3H9BF4O
Formula Weight: 147.91
Storage Temperature: -30°C to -10°C
Melting Point: ca 190° dec.
Beilstein Reference: 3597303
UN No.: 3261
MDL No.: MFCD00011798
Formula:
C₃H₉BF₄O MW: 147,91 g/mol Melting Pt: ∼ 190 °C (dec.) |
MDL Number:
MFCD00011798 CAS Number: 420-37-1 EINECS: 206-994-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...