2-Acetyl-7-methoxybenzofuran ≥97%

Supplier: Thermo Scientific

Synonyms: 1-(7-Methoxy-1-benzofuran-2-yl)-1-ethanone, 1-(7-Methoxybenzofuran-2-yl)ethanone

B21139.06 B21139.14
B21139.06EA 0 EUR
B21139.06 B21139.14
2-Acetyl-7-methoxybenzofuran ≥97%
2-Acetyl-7-methoxybenzofuran

Purity: 97% CAS No.: 43071-52-9 Molecular Formula: C11H10O3 Formula Weight: 190.20 Melting Point: 91-94° MDL No.: MFCD00016911

Formula: C₁₁H₁₀O₃
MW: 190,2 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00016911
CAS Number: 43071-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR